Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034812.2 | NP_001029984.1 | 1 - 2 | 196 | 115 | |
GenBank mRNA | BC102145.1 | - | 1 - 2 | 197 | 115 | |
BT021063.1 | - | 1 - 2 | 188 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |