Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035021.1 | NP_001030193.1 | 6 - 7 | 939 | 70 | |
GenBank mRNA | BC103359.1 | - | 6 - 7 | 939 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |