Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035024.1 | NP_001030196.1 | 8 - 9 | 707 | 66 | |
GenBank mRNA | BC102990.1 | - | 8 - 9 | 707 | 66 | |
BT026310.1 | - | 8 - 9 | 719 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |