Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035033.2 | NP_001030205.1 | 6 - 7 | 1214 | 70 | |
GenBank mRNA | BC102774.1 | - | 6 - 7 | 1215 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |