Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035034.2 | NP_001030206.1 | 5 - 6 | 419 | 89 | |
GenBank mRNA | BC102541.1 | - | 5 - 6 | 420 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |