Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035064.1 | NP_001030236.1 | 6 - 7 | 559 | 67 | |
GenBank mRNA | BC104588.1 | - | 6 - 7 | 559 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |