Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTAQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035091.1 | NP_001030263.1 | 4 - 5 | 530 | 126 | |
GenBank mRNA | BC102447.1 | - | 4 - 5 | 530 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |