Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035095.2 | NP_001030267.1 | 2 - 3 | 208 | 83 | |
GenBank mRNA | BC102817.1 | - | 2 - 3 | 209 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |