Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035293.1 | NP_001030370.1 | 2 - 3 | 1181 | 110 | |
GenBank mRNA | AY267257.1 | - | 1040 | 110 | ||
BC103349.1 | - | 2 - 3 | 1181 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |