Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035305.2 | NP_001030382.1 | 9 - 10 | 1453 | 77 | |
GenBank mRNA | BC107540.1 | - | 9 - 10 | 1454 | 77 | |
BT030747.1 | - | 9 - 10 | 1346 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |