Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035309.1 | NP_001030386.1 | 7 - 8 | 1014 | 105 | |
GenBank mRNA | BC105228.1 | - | 7 - 8 | 1014 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |