Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035337.1 | NP_001030414.1 | 3 - 4 | 474 | 72 | |
GenBank mRNA | BC102568.1 | - | 3 - 4 | 474 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |