Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035350.2 | NP_001030427.1 | 1 - 2 | 100 | 55 | |
GenBank mRNA | BC102534.1 | - | 1 - 2 | 102 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |