Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNJ2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035355.2 | NP_001030432.1 | 3 - 4 | 390 | 73 | |
GenBank mRNA | BC102451.1 | - | 3 - 4 | 391 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |