Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035419.2 | NP_001030496.1 | 5 - 6 | 487 | 74 | |
GenBank mRNA | BC102309.1 | - | 5 - 6 | 488 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |