Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035446.2 | NP_001030523.1 | 3 - 4 | 734 | 89 | |
GenBank mRNA | BC102550.1 | - | 3 - 4 | 735 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |