Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035452.2 | NP_001030529.1 | 3 - 4 | 276 | 64 | |
GenBank mRNA | BC103056.1 | - | 3 - 4 | 277 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |