Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035453.2 | NP_001030530.1 | 1 - 2 | 604 | 92 | |
GenBank mRNA | BC102688.1 | - | 1 - 2 | 605 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |