Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035471.1 | NP_001030548.1 | 3 - 4 | 368 | 94 | |
GenBank mRNA | BC103012.1 | - | 3 - 4 | 368 | 94 | |
BC151416.1 | - | 3 - 4 | 294 | 94 | ||
BT029883.1 | - | 3 - 4 | 321 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |