Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035501.1 | NP_001030578.1 | 5 - 6 | 713 | 74 | |
GenBank mRNA | BC103272.1 | - | 5 - 6 | 713 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |