Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037450.2 | NP_001032527.1 | 4 - 5 | 370 | 60 | |
GenBank mRNA | BC108102.1 | - | 4 - 5 | 371 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |