Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037462.2 | NP_001032539.1 | 2 - 3 | 183 | 127 | |
GenBank mRNA | BC108155.1 | - | 2 - 3 | 184 | 127 | |
BT025351.1 | - | 2 - 3 | 146 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |