Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037477.1 | NP_001032554.1 | 4 - 5 | 558 | 65 | |
GenBank mRNA | BC108103.1 | - | 4 - 5 | 558 | 65 | |
BT030492.1 | - | 4 - 5 | 571 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |