Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037479.2 | NP_001032556.1 | 4 - 5 | 1021 | 96 | |
GenBank mRNA | BC108111.1 | - | 4 - 5 | 1022 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |