Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037619.2 | NP_001032708.1 | 4 - 5 | 481 | 126 | |
GenBank mRNA | BC109831.1 | - | 4 - 5 | 482 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |