Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037818.1 | NP_001032907.1 | 4 - 5 | 283 | 63 | |
GenBank mRNA | BC107551.1 | - | 4 - 5 | 283 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |