Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038506.2 | NP_001033595.1 | 3 - 4 | 306 | 144 | |
GenBank mRNA | BC109775.1 | - | 3 - 4 | 307 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |