Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040472.2 | NP_001035562.2 | 5 - 6 | 538 | 64 | |
GenBank mRNA | BC103010.1 | - | 5 - 6 | 534 | 64 | |
DV802141.1 | - | 5 - 6 | 527 | 64 | ||
EH165860.1 | - | 5 - 6 | 539 | 64 | ||
EV719249.1 | - | 2 - 3 | 602 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |