Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040483.1 | NP_001035573.1 | 7 - 8 | 787 | 128 | |
GenBank mRNA | BC102067.1 | - | 7 - 8 | 787 | 128 | |
MK424004.1 | - | 1 - 2 | 101 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |