Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040581.2 | NP_001035671.1 | 157 | 57 | ||
GenBank mRNA | BC108180.1 | - | 158 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus |