Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045866.1 | NP_001039331.1 | 3 - 4 | 472 | 70 | |
GenBank mRNA | BC114643.1 | - | 3 - 4 | 1164 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |