Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075122.1 | NP_001068590.1 | 9 - 10 | 1202 | 70 | |
GenBank mRNA | BC105264.1 | - | 9 - 10 | 1202 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |