Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075494.2 | NP_001068962.1 | 1 - 2 | 59 | 90 | |
GenBank mRNA | BC120473.1 | - | 1 - 2 | 57 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |