Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075717.2 | NP_001069185.1 | 2 - 3 | 519 | 102 | |
GenBank mRNA | BC102601.1 | - | 2 - 3 | 521 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |