Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHHADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075780.2 | NP_001069248.2 | 1 - 2 | 111 | 113 | |
GenBank mRNA | BC102238.1 | - | 1 - 2 | 98 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |