Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075823.2 | NP_001069291.1 | 3 - 4 | 486 | 87 | |
GenBank mRNA | BC102208.1 | - | 3 - 4 | 488 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |