Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075933.1 | NP_001069401.1 | 2 - 3 | 494 | 70 | |
GenBank mRNA | BC102722.1 | - | 2 - 3 | 494 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |