Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076334.2 | NP_001069802.1 | 3 - 4 | 219 | 85 | |
GenBank mRNA | BC108201.1 | - | 3 - 4 | 220 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |