Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076461.2 | NP_001069929.1 | 4 - 5 | 362 | 121 | |
GenBank mRNA | BC102719.1 | - | 4 - 5 | 363 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |