Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077027.2 | NP_001070495.1 | 1 - 2 | 397 | 71 | |
GenBank mRNA | BC108229.1 | - | 1 - 2 | 401 | 71 | |
BC142383.1 | - | 1 - 2 | 307 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |