Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077045.2 | NP_001070513.1 | 3 - 4 | 453 | 98 | |
GenBank mRNA | BC103446.1 | - | 3 - 4 | 454 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |