Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077963.2 | NP_001071431.1 | 2 - 3 | 296 | 128 | |
GenBank mRNA | BC102714.1 | - | 2 - 3 | 297 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |