Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078037.2 | NP_001071505.1 | 3 - 4 | 259 | 99 | |
GenBank mRNA | BC102537.1 | - | 3 - 4 | 260 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |