Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL24D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080355.2 | NP_001073824.1 | 3 - 4 | 306 | 79 | |
GenBank mRNA | BC103265.1 | - | 3 - 4 | 308 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |