Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083640.1 | NP_001077109.1 | 7 - 8 | 1256 | 77 | |
GenBank mRNA | BC146260.1 | - | 7 - 8 | 1435 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |