Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083640.1 | NP_001077109.1 | 8 - 9 | 1349 | 74 | |
GenBank mRNA | BC146260.1 | - | 8 - 9 | 1528 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |