Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173945.4 | NP_776370.2 | 1 - 2 | 158 | 126 | |
GenBank mRNA | BC102381.1 | - | 1 - 2 | 159 | 126 | |
DQ515198.1 | - | 1 - 2 | 114 | 126 | ||
M18621.1 | - | 1 - 2 | 130 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |