Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173999.3 | NP_776424.1 | 1 - 2 | 460 | 83 | |
GenBank mRNA | AF014008.1 | - | 1 - 2 | 445 | 83 | |
BC102787.1 | - | 1 - 2 | 462 | 83 | ||
BC140655.1 | - | 1 - 2 | 461 | 83 | ||
BC142147.1 | - | 1 - 2 | 448 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |