Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174119.4 | NP_776544.1 | 9 - 10 | 936 | 79 | |
GenBank mRNA | AF079302.1 | - | 9 - 10 | 919 | 79 | |
BC103145.1 | - | 9 - 10 | 938 | 79 | ||
BT020852.1 | - | 9 - 10 | 952 | 79 | ||
BT021181.1 | - | 9 - 10 | 924 | 79 | ||
KF700639.1 | - | 10 - 11 | 1199 | 79 | ||
KF861986.1 | - | 8 - 9 | 1696 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |