Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174332.3 | NP_776757.1 | 1 - 2 | 612 | 104 | |
GenBank mRNA | AY049726.2 | - | 1 - 2 | 612 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |